tert-Butyl chloroacetate 97%

Supplier: Thermo Scientific Chemicals
Danger

34723-0250 34723-1000 34723-5000
TS34723-0250EA 35 USD
TS34723-0250 TS34723-1000 TS34723-5000
tert-Butyl chloroacetate 97%
tert-Butyl chloroacetate
Formula: C₆H₁₁ClO₂
Boiling Pt: 48…49 °C (11 mmHg)
Density: 1.053 g/cm³
Flash Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00000930
CAS Number: 107-59-5
EINECS: 203-506-1
UN: 2929
ADR: 6.1,II
Merck Index: 13,01562

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4220 to 1.4240 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR