Methyl bromoacetate 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Bromoacetic methyl ester

16955-0025 16955-0250 16955-1000 16955-5000
TS16955-0025EA 1109 USD
TS16955-0025 TS16955-0250 TS16955-1000 TS16955-5000
Methyl bromoacetate 99%
Methyl bromoacetate
Formula: BrCH₂COOCH₃
Boiling Pt: 145 °C (1013 hPa)
Melting Pt: –50 °C
Density: 1.61 g/cm³
Flash Pt: 62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000189
CAS Number: 96-32-2
EINECS: 202-499-2
UN: 2643
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4570 to 1.4590 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR