Benzyl bromoacetate 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzyl 2-bromoacetate, Bromoacetic benzyl ester

27804-0010 27804-0500 27804-2500
TS27804-0010EA 382 USD
TS27804-0010 TS27804-0500 TS27804-2500
Benzyl bromoacetate 96%
Benzyl bromoacetate
Formula: C₉H₉BrO₂
MW: 229.07 g/mol
Boiling Pt: 166…170 °C (22 mmHg)
Density: 1.44 g/cm³
Flash Pt: >109 °C
Storage Temperature: Ambient
MDL Number: MFCD00000190
CAS Number: 5437-45-6
EINECS: 226-611-4

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5430 to 1.5450 (20°C, 589 nm)
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR