Levulinic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Oxovaleric acid, 4-Ketovaleric acid, 4-Oxopentanoic acid

12514-0010 12514-0050 12514-1000 12514-2500
TS12514-0010EA 135 USD
TS12514-0010 TS12514-0050 TS12514-1000 TS12514-2500
Levulinic acid 98+%
Levulinic acid
Formula: C₅H₈O₃
MW: 116.12 g/mol
Boiling Pt: 245…246 °C (760 mmHg)
Melting Pt: 33…35 °C
Density: 1.13 g/cm³
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00002796
CAS Number: 123-76-2
EINECS: 204-649-2
Merck Index: 13,05492

Order Now

Specification Test Results

Appearance Colorless to white to yellow to orange Liquid after melting or solid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4390 to 1.4430 (20°C, 589 nm)
Titration with NaOH ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR