Methyl-p-toluate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Methylbenzoic acid methyl ester, Methyl-4-methylbenzoate, p-Toluic acid methyl ester, 4-Toluic acid methyl ester, Methyl 4-methylbenzoate

41496-0025 41496-0100 41496-1000 41496-2500
TS41496-0025EA 368 USD
TS41496-0025 TS41496-0100 TS41496-1000 TS41496-2500
Methyl-p-toluate 99%
Methyl-p-toluate
Formula: C₉H₁₀O₂
Boiling Pt: 220…223 °C
Melting Pt: 32…35 °C
Density: 1.06 g/cm³
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00008441
CAS Number: 99-75-2
EINECS: 202-784-1

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid or solid
Infrared spectrum Conforms
Melting point 32 to 35 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR