3,3'-Thiodipropionic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,3-Thiodipropionic acid

13875-0025 13875-0250 13875-5000
TS13875-0025EA 172 USD
TS13875-0025 TS13875-0050 TS13875-0250 TS13875-5000
3,3'-Thiodipropionic acid 99%
3,3'-Thiodipropionic acid
Formula: C₆H₁₀O₄S
MW: 178.21 g/mol
Melting Pt: 130…133 °C
Flash Pt: 128 °C
Storage Temperature: Ambient
MDL Number: MFCD00002781
CAS Number: 111-17-1
EINECS: 203-841-3
Merck Index: 12,09468

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 130 to 133 °C
Titration with NaOH 98.5 to 101.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR