N-Methylphthalimide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methylisoindoline-1,3-dione

30510-0050 30510-0250 30510-1000
TS30510-0050EA 26 USD
TS30510-0050 TS30510-0250 TS30510-1000
N-Methylphthalimide 98%
N-Methylphthalimide
Formula: C₉H₇NO₂
MW: 161.16 g/mol
Melting Pt: 129…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00023063
CAS Number: 550-44-7

Order Now

Specification Test Results

Appearance White to almost white Powder or crystals
Infrared spectrum Conforms
Melting point 132 to 136 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR