di-Sodium succinate hexahydrate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sodium succinate hexahydrate, Succinic acid disodium hexahydrate, 1,4-Butanedioic acid disodium salt hexahydrate, Sodium Succinate, Hexahydrate, Sodium succinate dibasic hexahydrate, Disodium succinate hexahydrate, Succinic acid disodium salt hexahydrate

15875-0010 15875-1000 15875-5000
TS15875-0010EA 181 USD
TS15875-0010 TS15875-1000 TS15875-5000
di-Sodium succinate hexahydrate 97%
di-Sodium succinate hexahydrate
Formula: NaOOCCH₂CH₂COONa·6H₂O
MW: 270.14 g/mol
Melting Pt: 300
Storage Temperature: Ambient
MDL Number: MFCD00149117
CAS Number: 6106-21-4
EINECS: 205-778-7
Merck Index: 13,08754

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals or granules
Infrared spectrum Conforms
Loss on drying 37.0 % to 41.0 % (90°C)
Solubility (10 % in water) Clear
Titration with HClO4 96.0 to 103.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR