Phthalimide 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Phthalic acid imide
13110-0010
13110-0025
13110-1000
13110-5000
TS13110-0010EA
77
USD
TS13110-0010
TS13110-0025
TS13110-1000
TS13110-5000
Phthalimide 99%
Phthalimide
Formula:
C₈H₅NO₂ Boiling Pt: 366 °C (760 mmHg) Melting Pt: >233.5 °C Flash Pt: 165 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005881 CAS Number: 85-41-6 EINECS: 201-603-3 Merck Index: 12,07529 |
Specification Test Results
Appearance | White to light yellow Powder or flakes |
Infrared spectrum | Conforms |
Melting point | 232 to 236 °C |
HPLC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...