Tetramethylurea 99%

Supplier: Thermo Scientific Chemicals
Danger

13849-0010 13849-1000 13849-2500
TS13849-0010EA 574 USD
TS13849-0010 TS13849-1000 TS13849-2500
Tetramethylurea 99%
Tetramethylurea
Formula: C₅H₁₂N₂O
Boiling Pt: 174…178 °C (760 mmHg)
Melting Pt: –1 °C
Density: 0.9879 g/cm³
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00008319
CAS Number: 632-22-4
EINECS: 211-173-9
Merck Index: 12,09368

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4490 to 1.4520 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR