1,1-Dimethylurea 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: N,N-Dimethylurea

38874-0050 38874-0250 38874-1000
TS38874-0050EA 32 USD
TS38874-0050 TS38874-0250 TS38874-1000
1,1-Dimethylurea 98+%
1,1-Dimethylurea
Formula: C₃H₈N₂O
MW: 88.11 g/mol
Melting Pt: 179…183 °C
Storage Temperature: Ambient
MDL Number: MFCD00007959
CAS Number: 598-94-7

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 179 to 183 °C
Titration with HClO4 ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR