p-Tolylacetic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
13912-0050
13912-0250
13912-1000
TS13912-0050EA
27
USD
TS13912-0050
TS13912-0250
TS13912-1000
p-Tolylacetic acid 99%
p-Tolylacetic acid
Formula:
(C₆H₄(CH₃))CH₂COOH Boiling Pt: 265…267 °C (760 mmHg) Melting Pt: 90…93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004353 CAS Number: 622-47-9 EINECS: 210-738-7 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 90 to 93 °C |
Titration with NaOH | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...