Ethyl decanoate 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: Ethyl caprate, Capric acid ethyl ester, Decanoic acid ethyl ester, Decylic acid ethyl ester

16500-0025 16500-0050 16500-1000 16500-5000
TS16500-0025EA 359 USD
TS16500-0025 TS16500-0050 TS16500-1000 TS16500-5000
Ethyl decanoate 99+%
Ethyl decanoate
Formula: C₁₂H₂₄O₂
MW: 200.32 g/mol
Boiling Pt: 245 °C (760 mmHg)
Melting Pt: –20 °C
Density: 0.8665 g/cm³
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00009581
CAS Number: 110-38-3
EINECS: 203-761-9
Merck Index: 13,03811

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4240 to 1.4270 (20°C, 589 nm)
Water ≤0.3 % (K.F.)
Density (20°C) 0.864 to 0.869 kg/l
GC ≥99.0 %
Acid value ≤1 mg KOH/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR