2-Cyclohexen-1-ol 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cyclohex-2-ene-1-ol, Cyclohex-2-enol

11123-0010 11123-0050 11123-0100
TS11123-0010EA 63 USD
TS11123-0010 TS11123-0050 TS11123-0100
2-Cyclohexen-1-ol 95%
2-Cyclohexen-1-ol
Formula: C₆H₁₀O
MW: 98.14 g/mol
Boiling Pt: 164…166 °C (760 mmHg)
Density: 0.984 g/cm³
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00001570
CAS Number: 822-67-3
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥94 %
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR