N,N-Dimethylformamide diethyl acetal 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,1-Diethoxy-N,N-dimethylmethanamine, 1,1-Diethoxytrimethylamine

16208-0100 16208-0500
TS16208-0100EA 35 USD
TS16208-0100 TS16208-0500
N,N-Dimethylformamide diethyl acetal 95%
N,N-Dimethylformamide diethyl acetal
Formula: C₇H₁₇NO₂
MW: 147.22 g/mol
Boiling Pt: 128…130 °C (760 mmHg)
Density: 0.859 g/cm³
Flash Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD00009227
CAS Number: 1188-33-6
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3990 to 1.4010 (20°C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR