(±)-4-Phenyl-2-butanol 97%
Supplier: Thermo Scientific Chemicals
18340-0050
18340-0250
TS18340-0050EA
36
USD
TS18340-0050
TS18340-0250
(±)-4-Phenyl-2-butanol 97%
(±)-4-Phenyl-2-butanol
Formula:
(C₆H₅)CH₂CH₂CH(OH)CH₃ MW: 150.22 g/mol Boiling Pt: 132 Density: 0.98 g/cm³ Flash Pt: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00044349 CAS Number: 2344-70-9 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5150 to 1.5180 (20°C, 589 nm) |
GC | ≥96.0 % |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...