Nicotinic acid (Niacin) 99.5%

Supplier: Thermo Scientific Chemicals
Warning

12829-0025 12829-0050 12829-0100 12829-1000 12829-5000
TS12829-0025EA 381 USD
TS12829-0025 TS12829-0050 TS12829-0100 TS12829-1000 TS12829-5000
Nicotinic acid (Niacin) 99.5%
Nicotinic acid (Niacin)
Formula: C₆H₅NO₂
MW: 123.11 g/mol
Melting Pt: 236…239 °C
Storage Temperature: Ambient
MDL Number: MFCD00006391
CAS Number: 59-67-6
EINECS: 200-441-0
Merck Index: 13,06552

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 234 to 240 °C
Loss on drying ≤1 % (105 °C, 1 h)
Sulfated ash ≤0.1 %
Titration with NaOH ≥99.4 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR