1-Methylcyclopropanecarboxylic acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
19496-0010
19496-0050
TS19496-0010EA
67
USD
TS19496-0010
TS19496-0050
1-Methylcyclopropanecarboxylic acid 98%
1-Methylcyclopropanecarboxylic acid
Formula:
C₅H₈O₂ Melting Pt: 34…36 °C Flash Pt: 84 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001290 CAS Number: 6914-76-7 UN: 3261 ADR: 8,II |
Specification Test Results
Appearance | White to light yellow Low melting solid |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...