2,3-Dimethyl-1,3-butadiene 98% stabilized

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,3-Dimethylbuta-1,3-diene

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

16343-0050 16343-0250 16343-1000
TS16343-0050EA 47 USD
TS16343-0050 TS16343-0250 TS16343-1000
2,3-Dimethyl-1,3-butadiene 98% stabilized
2,3-Dimethyl-1,3-butadiene
Formula: C₆H₁₀
MW: 82.15 g/mol
Boiling Pt: 68…69 °C (760 mmHg)
Melting Pt: –76 °C
Density: 0.726 g/cm³
Flash Pt: –20 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008595
CAS Number: 513-81-5
UN: 3295
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4370 to 1.4400 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer 100 ppm BHT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR