Triphenyl phosphate 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Triphenylphosphate, TPP

14767-0010 14767-0025 14767-0050 14767-2500
TS14767-0010EA 74 USD
TS14767-0010 TS14767-0025 TS14767-0050 TS14767-2500
Triphenyl phosphate 99+%
Triphenyl phosphate
Formula: C₁₈H₁₅O₄P
MW: 326.29 g/mol
Boiling Pt: 412
Melting Pt: 47…53 °C
Flash Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00003031
CAS Number: 115-86-6
EINECS: 204-112-2
UN: 3077
ADR: 9 III
Merck Index: 12,09872

Order Now

Specification Test Results

Appearance White Flakes and/or pastilles
Infrared spectrum Conforms
Melting point 47 to 53 °C
Water ≤0.1 %
Additional info Free phenols: < 0.005%
GC ≥99.0 %
Color scale ≤50 APHA (after heat treatment)
Acid value ≤0.05 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR