Diammonium hydrogen citrate 99+% for biochemistry

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Citric acid diammonium salt, Diammonium hydrogen 2-hydroxypropane-1,2,3-tricarboxylate, Ammonium citrate dibasic

44802-1000 44802-5000
TS44802-1000EA 50 USD
TS44802-1000 TS44802-5000
Diammonium hydrogen citrate 99+% for biochemistry
Diammonium hydrogen citrate
Formula: HOC(CO₂H)(CH₂CO₂NH₄)₂
MW: 226.19 g/mol
Density: 1.5 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00013068
CAS Number: 3012-65-5
Merck Index: 13,00513

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Solubility Clear colorless
UV at 280 nm A: ≤0.1
Titration with HClO4 ≥99.0 %
Chloride (Cl) ≤0.001 %
Iron (Fe) ≤0.001 %
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR