1-Methylcyclohexene 98+% stabilized

Supplier: Thermo Scientific Chemicals
Danger

15684-0100 15684-0500
TS15684-0100EA 64 USD
TS15684-0100 TS15684-0500
1-Methylcyclohexene 98+% stabilized
1-Methylcyclohexene
Formula: C₇H₁₂
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: –120.4 °C
Density: 0.81 g/cm³
Flash Pt: –3 °C
Storage Temperature: Ambient
MDL Number: MFCD00001548
CAS Number: 591-49-1
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4492 to 1.4512 (20°C, 589 nm)
GC ≥98.0 %
Stabilizer 10 ppm hydroquinone
Sulfate (SO4) ≤0.015 %
Ammonium (NH4) ≤0.001 %
Iron (Fe) ≤0.001 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR