5-Quinolinol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Hydroxyquinoline, Quinolin-5-ol

15643-0010 15643-0050
TS15643-0010EA 87 USD
TS15643-0010 TS15643-0050
5-Quinolinol 97%
5-Quinolinol
Formula: C₉H₇NO
MW: 145.16 g/mol
Melting Pt: 223…226 °C
Storage Temperature: Ambient
MDL Number: MFCD00006792
CAS Number: 578-67-6

Order Now

Specification Test Results

Appearance White to yellow to beige to brown Powder or crystals
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR