2-Octyne 97% stabilized

Supplier: Thermo Scientific Chemicals
Danger

45557-0050 45557-0250
TS45557-0050EA 30 USD
TS45557-0050 TS45557-0250
2-Octyne 97% stabilized
2-Octyne
Formula: C₈H₁₄
MW: 110.2 g/mol
Boiling Pt: 137 °C (760 mmHg)
Density: 0.759 g/cm³
Flash Pt: 18 °C
Storage Temperature: Ambient
MDL Number: MFCD00027302
CAS Number: 2809-67-8
UN: 3295
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4270 to 1.4300 (20°C, 589 nm)
GC ≥96.0 %
Stabilizer Approx. 200 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR