Citric acid monohydrate 99+% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid
38585-0010
38585-0025
38585-2500
TS38585-0010EA
68
USD
TS38585-0010
TS38585-0025
TS38585-2500
Citric acid monohydrate 99+% ACS
Citric acid monohydrate
Formula:
C₆H₈O₇.H₂O MW: 210.14 g/mol Melting Pt: 135…152 °C Flash Pt: 174 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149972 CAS Number: 5949-29-1 Merck Index: 13,02350 |
|
Specification Test Results
Appearance | Colorless to white Powder or crystals |
Residue after ignition | ≤0.02 % |
Titration with NaOH | 99.0 to 102.0 % |
Carboniz. sub. by hot H2SO4 | Passes test |
Chloride (Cl) | ≤0.001 % |
Oxalate (C2O4) | (limit about 0.05%) Passes test |
Phosphate (PO4) | ≤0.001 % |
Sulfate (SO4) | ≤0.002 % |
Iron (Fe) | ≤3 ppm |
Lead (Pb) | ≤2 ppm |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...