2-Fluorocinnamic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(2-Fluorophenyl)propenoic acid

20710-0250 207100010 207100050
TS20710-0250EA 169 USD
TS20710-0250 TS207100010 TS207100050
2-Fluorocinnamic acid 98%
2-Fluorocinnamic acid
Formula: C₉H₇FO₂
MW: 166.15 g/mol
Melting Pt: 179…183 °C
Storage Temperature: Ambient
MDL Number: MFCD00004370
CAS Number: 451-69-4
EINECS: 207-195-3

Order Now

Specification Test Results

Appearance White Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 179 to 183 °C
HPLC ≥97.5 %
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR