4-Fluorocinnamic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2E)-3-(4-fluorophenyl)prop-2-enoic acid, p-Fluorocinnamic acid, 3-4-fluorophenyl acrylic acid, trans-4-Fluorocinnamic acid

20711-0050 20711-0250
TS20711-0050EA 41 USD
TS20711-0050 TS20711-0250
4-Fluorocinnamic acid 99%
4-Fluorocinnamic acid
Formula: C₉H₇FO₂
MW: 166.15 g/mol
Melting Pt: 207…210 °C
MDL Number: MFCD00004395
CAS Number: 459-32-5
EINECS: 207-288-9

Order Now

Specification Test Results

Appearance White to off-white Crystalline solid
Infrared spectrum Conforms
Melting point 207 to 210 °C
HPLC ≥98.5 %
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR