Dodecylbenzene sulfonic acid mixture of C10-C13 isomers

Supplier: Thermo Scientific Chemicals
Danger

32590-0010 32590-5000
TS32590-0010EA 325 USD
TS32590-0010 TS32590-5000
Dodecylbenzene sulfonic acid mixture of C10-C13 isomers
Dodecylbenzene sulfonic acid mixture of C10-C13 isomers
Formula: C₁₈H₃₀O₃S
Boiling Pt: 315 °C (1013 hPa)
Melting Pt: –10 °C
Density: 1.06 g/cm³
Flash Pt: 210 °C
MDL Number: MFCD00066677
CAS Number: 85536-14-7
EINECS: 248-289-4
UN: 2586
ADR: 8,III

Order Now

Specification Test Results

Appearance Brown Viscous liquid
Infrared spectrum Conforms
Assay ≥96 % (Sulfonic acid, M.W. 322)
Water ≤0.5 % (K.F.)
Impurity ≤3 % Neutral oil
Sulfuric acid ≤1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR