4-Methoxycinnamic acid predominantly trans 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(4-Methoxyphenyl)acrylic acid

12584-0250 12584-1000
TS12584-0250EA 62 USD
TS12584-0250 TS12584-1000
4-Methoxycinnamic acid predominantly trans 99%
4-Methoxycinnamic acid
Formula: C₁₀H₁₀O₃
MW: 178.18 g/mol
Melting Pt: 173…175 °C
MDL Number: MFCD00004398
CAS Number: 830-09-1
EINECS: 212-594-0

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 173 to 175 °C
Titration with NaOH ≥98.5 %
Chloride (Cl) ≤0.0001 %
Sulfide (S) ≤0.00001 %
Phosphate (PO4) ≤0.0001 %
Carbonate (as CO2) ≤0.002 %
Aluminium (Al) ≤0.00001 %
Antimony (Sb) ≤0.000002 %
Arsenic (As) ≤0.00001 %
Barium (Ba) ≤0.00001 %
Beryllium (Be) ≤0.00001 %
Bismuth (Bi) ≤0.000005 %
Cadmium (Cd) ≤0.00001 %
Calcium (Ca) ≤0.0001 %
Chromium (Cr) ≤0.000005 %
Cobalt (Co) ≤0.00001 %
Gallium (Ga) ≤0.000005 %
Germanium (Ge) ≤0.000002 %
Gold (Au) ≤0.00001 %
Indium (In) ≤0.000005 %
Iron (Fe) ≤0.00001 %
Lead (Pb) ≤0.000005 %
Lithium (Li) ≤0.000002 %
Magnesium (Mg) ≤0.00005 %
Manganese (Mn) ≤0.000001 %
Mercury (Hg) ≤0.00001 %
Molybdenum (Mo) ≤0.000002 %
Nickel (Ni) ≤0.000005 %
Platinum (Pt) ≤0.00001 %
Potassium (K) ≤0.0001 %
Silicon (Si) ≤0.00002 %
Silver (Ag) ≤0.000002 %
Sodium (Na) ≤0.0002 %
Strontium (Sr) ≤0.0001 %
Thallium (Tl) ≤0.000002 %
Tin (Sn) ≤0.000005 %
Titanium (Ti) ≤0.000005 %
Vanadium (V) ≤0.000002 %
Zinc (Zn) ≤0.00001 %
Zirconium (Zr) ≤0.000005 %
Heavy metals (as Pb) ≤1 ppm
KMnO4-reducing substance ≤0.001 % (as O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR