Triethyl-2-phosphonobutyrate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 2-(diethoxyphosphoryl)butanoate

36878-0050 36878-0250
TS36878-0050EA 27 USD
TS36878-0050 TS36878-0250
Triethyl-2-phosphonobutyrate 98%
Triethyl 2-phosphonobutyrate
Formula: C₁₀H₂₁O₅P
MW: 252.25 g/mol
Boiling Pt: 152…154 °C (14 mmHg)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00041347
CAS Number: 17145-91-4

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4315 to 1.4335 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR