N,N-Diisopropylcarbamoyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Diisopropylcarbamoyl Chloride

31452-0050 31452-0250
TS31452-0050EA 61 USD
TS31452-0050 TS31452-0250
N,N-Diisopropylcarbamoyl chloride 98%
N,N-Diisopropylcarbamoyl chloride
Formula: C₇H₁₄ClNO
Boiling Pt: 90…93 °C (15 mmHg)
Melting Pt: 58.1…59.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00015013
CAS Number: 19009-39-3
EINECS: 242-743-5
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to white to light yellow Crystalline powder or low melting solid
Infrared spectrum Conforms
Melting point 55 to 60 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR