Nicotinic acid hydrazide 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Nicotinohydrazide, (3-Pyridylcarbonyl)hydrazine, Pyridine-3-carbohydrazide

14820-0250 14820-1000
TS14820-0250EA 47 USD
TS14820-0250 TS14820-1000
Nicotinic acid hydrazide 97%
Nicotinic acid hydrazide
Formula: C₆H₇N₃O
Melting Pt: 160…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00006383
CAS Number: 553-53-7
EINECS: 209-041-0

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or needles
Infrared spectrum Conforms
Melting point 160 to 163 °C
Assay ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR