(-)-Dibenzoyl-L-tartaric acid monohydrate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(2R,3R)-(-)-Di-O-benzoyl-tartaric acid monohydrate
16025-1000
16025-5000
TS16025-1000EA
46
USD
TS16025-1000
TS16025-5000
(-)-Dibenzoyl-L-tartaric acid monohydrate 98%
(-)-Dibenzoyl-L-tartaric acid monohydrate
Formula:
C₁₈H₁₄O₈·1H₂O Melting Pt: 88…90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149119 CAS Number: 62708-56-9 EINECS: 220-374-0 |
Specification Test Results
Appearance | White Powder |
Infrared spectrum | Conforms |
Water | 4 to 5.5 % |
HPLC | ≥97.5 % |
Specific optical rotation | -113° to -117° (20°C, 589 nm) (c=5, ethanol) |
Titration with TBAH | 97.5 to 102.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...