Naphthol AS-BI phosphate 93%

Supplier: Thermo Scientific Chemicals
Warning

41531-0010 41531-1000
TS41531-0010EA 243 USD
TS41531-0010 TS41531-1000
Naphthol AS-BI phosphate 93%
Naphthol AS-BI phosphate
Formula: C₁₈H₁₅BrNO₆P
Storage Temperature: Refrigerator
MDL Number: MFCD00004058
CAS Number: 1919-91-1
EINECS: 217-645-0

Order Now

Specification Test Results

Appearance Light yellow Crystalline powder
Infrared spectrum Conforms
HPLC ≥91 %
GC ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR