Isethionic acid sodium salt (sodium 2-hydroxyethanesulfonate) 98%

Supplier: Thermo Scientific Chemicals

20500-0010 20500-5000
TS20500-0010EA 79 USD
TS20500-0010 TS20500-5000
Isethionic acid sodium salt (sodium 2-hydroxyethanesulfonate) 98%
Isethionic acid sodium salt (sodium 2-hydroxyethanesulphonate)
Formula: C₂H₅NaO₄S
Melting Pt: 191…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00007534
CAS Number: 1562-00-1
EINECS: 216-343-6

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 191 to 195 °C
Titration after Ion exchange ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR