Cyclopentylacetic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Cyclopentylacetic acid

15487-0100 15487-0250
TS15487-0100EA 84 USD
TS15487-0100 TS15487-0250
Cyclopentylacetic acid 97%
Cyclopentylacetic acid
Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Boiling Pt: 133…134 °C (23 mmHg)
Density: 1.02 g/cm³
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00001387
CAS Number: 1123-00-8
EINECS: 214-368-7

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4520 to 1.4540 (20°C, 589 nm)
Titration with NaOH ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR