S-Methylisothiouronium sulfate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: S-Methylisothiourea sulphate, 2-Methyl-2-thiopseudourea sulphate, S-Methylthiouronium sulphate, 2-Methyl-2-thiopseudourea hemisulphate

12789-1000 12789-5000
TS12789-1000EA 37 USD
TS12789-1000 TS12789-5000
S-Methylisothiouronium sulfate 98%
S-Methylisothiouronium sulphate
Formula: C₄H₁₂N₄S₂·H₂SO₄
MW: 278.38 g/mol
Melting Pt: 240…241 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00013055
CAS Number: 867-44-7
EINECS: 212-759-7

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR