Abietic acid 85%

Supplier: Thermo Scientific Chemicals
Warning

17313-0250 17313-1000
TS17313-0250EA 91 USD
TS17313-0250 TS17313-1000
Abietic acid 85%
Abietic acid
Formula: C₂₀H₃₀O₂
Storage Temperature: Ambient
MDL Number: MFCD03423567
CAS Number: 514-10-3
EINECS: 208-178-3
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to yellow Powder or crystals
Infrared spectrum Conforms
GC ≥80 %
Free fructose, glucose,sucrose ≤5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR