Triethyl-1,1,2-ethanetricarboxylate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Triethyl ethane-1,1,2-tricarboxylate

13961-0250 13961-1000
TS13961-0250EA 38 USD
TS13961-0250 TS13961-1000
Triethyl-1,1,2-ethanetricarboxylate 99%
Triethyl-1,1,2-ethanetricarboxylate
Formula: C₁₁H₁₈O₆
MW: 246.26 g/mol
Boiling Pt: 99 °C (.5 mmHg)
Density: 1.07 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00009154
CAS Number: 7459-46-3
EINECS: 231-235-9

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4280 to 1.4300 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR