N-(2-Chloroethyl)acetamide 98%

Supplier: Thermo Scientific Chemicals
Warning

18452-0100 18452-0500
TS18452-0100EA 51 USD
TS18452-0100 TS18452-0500
N-(2-Chloroethyl)acetamide 98%
N-(2-Chloroethyl)acetamide
Formula: C₄H₈ClNO
Density: 1.2 g/cm³
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000965
CAS Number: 7355-58-0
EINECS: 230-884-5

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4770 to 1.4800 (20°C, 589 nm)
GC ≥97.5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR