Benzyl carbamate 98%

Supplier: Thermo Scientific Chemicals

10589-0250 10589-1000
TS10589-0250EA 43 USD
TS10589-0250 TS10589-1000
Benzyl carbamate 98%
Benzyl carbamate
Formula: C₈H₉NO₂
MW: 151.17 g/mol
Melting Pt: 86…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00007965
CAS Number: 621-84-1
EINECS: 210-710-4

Order Now

Specification Test Results

Appearance Off-white to light beige Powder or flakes
Infrared spectrum Conforms
Melting point 86 to 89 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR