(S)-(+)-4-Phenyl-2-oxazolidinone 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (S)-4-Phenyloxazolidin-2-one

30385-0010 30385-0050
TS30385-0010EA 71 USD
TS30385-0010 TS30385-0050
(S)-(+)-4-Phenyl-2-oxazolidinone 99%
(S)-(+)-4-Phenyl-2-oxazolidinone
Formula: C₉H₉NO₂
MW: 163.18 g/mol
Melting Pt: 130…133 °C
Storage Temperature: Ambient
MDL Number: MFCD00043396
CAS Number: 99395-88-7

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 130 to 133 °C
GC ≥98.5 %
Specific optical rotation +48° to +51° (20 °C, 589 nm) (c=2, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR