4-Methoxy-2(5H)-furanone 97%

Supplier: Thermo Scientific Chemicals
Warning

27668-0050 27668-0250
TS27668-0050EA 57 USD
TS27668-0050 TS27668-0250
4-Methoxy-2(5H)-furanone 97%
4-Methoxy-2(5H)-furanone
Formula: C₅H₆O₃
MW: 114.1 g/mol
Boiling Pt: 130…131 °C (18mbar)
Melting Pt: 60…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00071565
CAS Number: 69556-70-3

Order Now

Specification Test Results

Appearance Light yellow Crystalline mass
Infrared spectrum Conforms
Melting point 60 to 65 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR