2-Chloro-N-(2,6-dimethylphenyl)acetamide 98%

Supplier: Thermo Scientific Chemicals
Warning

13544-0100 13544-0500
TS13544-0100EA 75 USD
TS13544-0100 TS13544-0500
2-Chloro-N-(2,6-dimethylphenyl)acetamide 98%
2-Chloro-N-(2,6-dimethylphenyl)acetamide
Formula: C₁₀H₁₂ClNO
Melting Pt: 143…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00000926
CAS Number: 1131-01-7
EINECS: 214-460-7

Order Now

Specification Test Results

Appearance White to brown Crystalline powder or needles
Infrared spectrum Conforms
Melting point 143 to 150 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR