2,2,6-Trimethyl-4H-1,3-dioxin-4-one 94%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,2,6-Trimethyl-1,3-dioxine-4-one

29691-1000 29691-5000
TS29691-1000EA 87 USD
TS29691-1000 TS29691-5000
2,2,6-Trimethyl-4H-1,3-dioxin-4-one 94%
2,2,6-Trimethyl-4H-1,3-dioxin-4-one
Formula: C₇H₁₀O₃
MW: 142.15 g/mol
Boiling Pt: 65…67 °C (2.7mbar)
Melting Pt: 4…13 °C
Density: 1.08 g/cm³
Flash Pt: 14 °C
Storage Temperature: Ambient
MDL Number: MFCD00040468
CAS Number: 5394-63-8
EINECS: 226-403-3
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear yellow to dark brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4580 to 1.4650 (20°C, 589 nm)
GC ≥92 %
Acetone ≤6 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR