Aluminon

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: ALuminon, Ammonium auritricarboxylate ATA Triammonium aurinetricarboxylate triammonium salt Ammonium aurintricarboxylate aurintricarboxylic acid ammonium salt Aluminon(C.1.No.43810) aurintricarboxylic acid triammonium salt, Ammonium 5,5'-((3-carboxylato-4-oxocyclohexa-2,5-dien-1-ylidene)methylene)bis(2-hydroxybenzoate), Aurin Tricarboxylic Acid Ammonium Salt

10306-0250 10306-1000
TS10306-0250EA 59 USD
TS10306-0250 TS10306-1000
Aluminon
Aluminon
Formula: C₂₂H₂₃N₃O₉
MW: 473.44 g/mol
Melting Pt: 220…225 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00040925
CAS Number: 569-58-4
EINECS: 209-319-1

Order Now

Specification Test Results

Appearance Red to yellow-brown to red-brown Crystalline powder
Infrared spectrum Conforms
Sulfated ash ≤0.2 %
Solubility (1.0 % in water) Clear
Suitability (for Al determination) Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR