Coumarin-3-carboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

34676-0250 34676-1000
TS34676-0250EA 71.5 USD
TS34676-0250 TS34676-1000
Coumarin-3-carboxylic acid 98%
Coumarin-3-carboxylic acid
Formula: C₁₀H₆O₄
Melting Pt: 190…193 °C
Storage Temperature: Ambient
MDL Number: MFCD00006852
CAS Number: 531-81-7
EINECS: 208-518-0
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Light brown Crystalline powder
Infrared spectrum Conforms
Titration with TBAH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR