Methyl oxalyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methyl 2-chloro-2-oxoacetate, Methyl chloroglyoxylate

16600-0250 16600-0500
TS16600-0250EA 129 USD
TS16600-0250 TS16600-0500
Methyl oxalyl chloride 97%
Methyl oxalyl chloride
Formula: C₃H₃ClO₃
MW: 122.51 g/mol
Boiling Pt: 118…120 °C (760 mmHg)
Density: 1.33 g/cm³
Flash Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00000705
CAS Number: 5781-53-3
EINECS: 227-307-4
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4179 to 1.4199 (20°C, 589 nm)
GC ≥96.0 %
Color scale ≤200 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR