N,N-Dimethylacetamide dimethyl acetal 97% stabilised

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (1,1-Dimethoxyethyl)dimethylamine, 1,1-Dimethoxy-N,N-dimethylethanamine

26413-0250 26413-1000
TS26413-0250EA 185 USD
TS26413-0250 TS26413-1000
N,N-Dimethylacetamide dimethyl acetal 97% stabilised
N,N-Dimethylacetamide dimethyl acetal
Formula: C₆H₁₅NO₂
Boiling Pt: 110…118 °C (760 mmHg)
Density: 0.911 g/cm³
Flash Pt: 8 °C
Storage Temperature: Ambient
MDL Number: MFCD00008476
CAS Number: 18871-66-4
EINECS: 242-641-0
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4100 to 1.4180 (20°C, 589 nm)
Stabilizer sodium carbonate
Titration with HClO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR