Indium(III) trifluoromethanesulphonate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Indium(III) trifluoromethanesulphonate, Indium triflate

36854-0050 36854-0250
TS36854-0050EA 111 USD
TS36854-0050 TS36854-0250
Indium(III) trifluoromethanesulphonate 98%
Indium(III) trifluoromethanesulphonate
Formula: C₃F₉InO₉S₃
Storage Temperature: Ambient
MDL Number: MFCD00144478
CAS Number: 128008-30-0
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to almost white Powder and/or granules
Infrared spectrum Conforms
Titration Complexometric 19.9 to 20.9 % (In)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR